Methyl 2-amino-4-bromo-6-fluorobenzoate

Lieferant: Apollo Scientific
Achtung

PC400191-1G PC400191-250MG PC400191-5G
APOSPC400191-1GEA 74.1 EUR
APOSPC400191-1G APOSPC400191-250MG APOSPC400191-5G
Methyl 2-amino-4-bromo-6-fluorobenzoate
Methyl 2-amino-4-bromo-6-fluorobenzoate
Formel: C₈H₇BrFNO₂
Molecular Weight: 248,049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28657035
CAS-Nummer: 1698028-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR