5-Amino-6-chloro-4-fluoro-1H-benzimidazole
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-4-fluoro-1H-benzimidazol-5-amine
PC400519-250MG
PC400519-500MG
APOSPC400519-250MGEA
524
EUR
APOSPC400519-250MG
APOSPC400519-500MG
5-Amino-6-chloro-4-fluoro-1H-benzimidazole
5-Amino-6-chloro-4-fluoro-1H-benzimidazole
Formel:
C₇H₅ClFN₃ Molecular Weight: 185,586 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22690795 CAS-Nummer: 1426290-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...