2,4,6-Trichloro-5-fluoropyrimidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4,6-Trichloro-5-fluoro-1,3-diazine
PC400523-1G
PC400523-5G
APOSPC400523-1GEA
126
EUR
APOSPC400523-1G
APOSPC400523-5G
2,4,6-Trichloro-5-fluoropyrimidine 97%
2,4,6-Trichloro-5-fluoropyrimidine
Formel:
C₄Cl₃FN₂ Molecular Weight: 201,414 g/mol Schmelzpunkt: 37…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040435 CAS-Nummer: 6693-08-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...