1-Benzyl-3,3-difluoropiperidin-4-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Benzyl-3,3-difluoro-4-oxopiperidine
PC400527-1G
PC400527-5G
APOSPC400527-1GEA
109
EUR
APOSPC400527-1G
APOSPC400527-5G
1-Benzyl-3,3-difluoropiperidin-4-one 95%
1-Benzyl-3,3-difluoropiperidin-4-one
Formel:
C₁₂H₁₃F₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD15526740 CAS-Nummer: 1039741-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...