2-Amino-4-(difluoromethyl)pyridine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Difluoromethyl)pyridin-2-amine

PC400579-1G PC400579-500MG PC400579-5G
APOSPC400579-1GEA 272 EUR
APOSPC400579-1G APOSPC400579-500MG APOSPC400579-5G
2-Amino-4-(difluoromethyl)pyridine 98%
2-Amino-4-(difluoromethyl)pyridine
Formel: C₆H₆F₂N₂
Molecular Weight: 144,122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25476528
CAS-Nummer: 1346536-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR