Ethyl 5-[3-(trifluoromethyl)phenyl]-1,3,4-oxadiazole-2-carboxylate
Lieferant: Apollo Scientific
Synonyme:
5-[3-(Trifluoromethyl)phenyl]-1,3,4-oxadiazole-2-carboxylic acid ethyl ester
PC400667-1G
PC400667-250MG
PC400667-500MG
PC400667-5G
APOSPC400667-1GEA
209
EUR
APOSPC400667-1G
APOSPC400667-250MG
APOSPC400667-500MG
APOSPC400667-5G
Ethyl 5-[3-(trifluoromethyl)phenyl]-1,3,4-oxadiazole-2-carboxylate
Ethyl 5-[3-(trifluoromethyl)phenyl]-1,3,4-oxadiazole-2-carboxylate
Formel:
C₁₂H₉F₃N₂O₃ Molecular Weight: 286,207 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24369730 CAS-Nummer: 1432053-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...