Benzyl-3-fluoro-4-oxopiperidine-1-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzyl 3-fluoro-4-oxopiperidine-1-carboxylate
PC400706-1G
PC400706-5G
APOSPC400706-1GEA
87
EUR
APOSPC400706-1G
APOSPC400706-5G
Benzyl-3-fluoro-4-oxopiperidine-1-carboxylate 95%
Benzyl-3-fluoro-4-oxopiperidine-1-carboxylate
Formel:
C₁₃H₁₄FNO₃ Molecular Weight: 251,254 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11976639 CAS-Nummer: 845256-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...