2-Acetyl-3-chloro-5-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-2-ethanoyl-5-(trifluoromethyl)pyridine, 1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethan-1-one
PC401010-1G
PC401010-500MG
PC401010-5G
APOSPC401010-1GEA
412
EUR
APOSPC401010-1G
APOSPC401010-500MG
APOSPC401010-5G
2-Acetyl-3-chloro-5-(trifluoromethyl)pyridine
2-Acetyl-3-chloro-5-(trifluoromethyl)pyridine
Formel:
C₈H₅ClF₃NO Molecular Weight: 223,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11113406 CAS-Nummer: 207994-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...