2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]propionic acid

Lieferant: Apollo Scientific

PC401030-1G PC401030-5G
APOSPC401030-1GEA 364 EUR
APOSPC401030-1G APOSPC401030-5G
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]propionic acid
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]propionic acid
Formel: C₉H₇ClF₃NO₂
Molecular Weight: 253,606 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26912299
CAS-Nummer: 1566079-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR