tert-Butyl 6-fluoro-4-iodo-1H-benzimidazole-1-carboxylate

Lieferant: Apollo Scientific

PC402001-1G PC402001-250MG
APOSPC402001-1GEA 669 EUR
APOSPC402001-1G APOSPC402001-250MG
tert-Butyl 6-fluoro-4-iodo-1H-benzimidazole-1-carboxylate
tert-Butyl 6-fluoro-4-iodo-1H-benzimidazole-1-carboxylate
Formel: C₁₂H₁₂FIN₂O₂
Molecular Weight: 362,139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13182503
CAS-Nummer: 895535-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR