5-Bromo-7-(trifluoromethyl)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC402014-250MG PC402014-100MG
APOSPC402014-250MGEA 85.7 EUR
APOSPC402014-250MG APOSPC402014-100MG
5-Bromo-7-(trifluoromethyl)-1H-indazole
5-Bromo-7-(trifluoromethyl)-1H-indazole
Formel: C₈H₄BrF₃N₂
Molecular Weight: 265,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21602947
CAS-Nummer: 1374258-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR