2-Bromo-1-(4-(fluoromethyl)phenyl)ethan-1-one 97%

Lieferant: Apollo Scientific

PC402128-1G PC402128-500MG
APOSPC402128-1GEA 5240 EUR
APOSPC402128-1G APOSPC402128-500MG
2-Bromo-1-(4-(fluoromethyl)phenyl)ethan-1-one 97%
2-Bromo-1-(4-(fluoromethyl)phenyl)ethan-1-one
Formel: C₉H₈BrFO
Molecular Weight: 231.062 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22492677
CAS-Nummer: 1628010-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR