7-Fluoro-4-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific

Synonyme: 7-Fluoro-4-hydroxy-2-methyl-1-azanaphthalene, 7-Fluoro-2-methylquinolin-4-ol

PC404503-1G PC404503-5G
APOSPC404503-1GEA 335 EUR
APOSPC404503-1G APOSPC404503-5G
7-Fluoro-4-hydroxy-2-methylquinoline
7-Fluoro-4-hydroxy-2-methylquinoline
Formel: C₁₀H₈FNO
Molecular Weight: 177,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684201
CAS-Nummer: 18529-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR