4-Amino-8-fluoro-2-methylquinoline

Lieferant: Apollo Scientific

Synonyme: 8-Fluoro-2-methylquinolin-4-amine, 4-Amino-8-fluoro-2-methyl-1-azanaphthalene

PC404504-1G PC404504-5G
APOSPC404504-1GEA 343 EUR
APOSPC404504-1G APOSPC404504-5G
4-Amino-8-fluoro-2-methylquinoline
4-Amino-8-fluoro-2-methylquinoline
Formel: C₁₀H₉FN₂
Molecular Weight: 176,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787728
CAS-Nummer: 288151-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR