4-(5-Fluoro-2-methyl-benzyl)-piperidine
Lieferant: Apollo Scientific
PC404523-1G
PC404523-250MG
APOSPC404523-1GEA
0
EUR
APOSPC404523-1G
APOSPC404523-250MG
4-(5-Fluoro-2-methyl-benzyl)-piperidine
4-(5-Fluoro-2-methyl-benzyl)-piperidine
Formel:
C₁₃H₁₈FN Molecular Weight: 207.287 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863585 CAS-Nummer: 782504-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...