4-Amino-8-fluoroquinoline

Lieferant: Apollo Scientific

PC404562-1G PC404562-5G
APOSPC404562-1GEA 426 EUR
APOSPC404562-1G APOSPC404562-5G
4-Amino-8-fluoroquinoline
4-Amino-8-fluoroquinoline
Formel: C₉H₇FN₂
Molecular Weight: 162,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04037191
CAS-Nummer: 148401-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR