4-Bromo-7,8-difluoroquinoline

Lieferant: Apollo Scientific

PC404584-1G PC404584-5G
APOSPC404584-1GEA 227 EUR
APOSPC404584-1G APOSPC404584-5G
4-Bromo-7,8-difluoroquinoline
4-Bromo-7,8-difluoroquinoline
Formel: C₉H₄BrF₂N
Molecular Weight: 244.036 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674974
CAS-Nummer: 1189106-43-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR