4-Chloro-6,8-difluoro-2-methylquinoline
Lieferant: Apollo Scientific
PC404589-1G
PC404589-5G
APOSPC404589-1GEA
343
EUR
APOSPC404589-1G
APOSPC404589-5G
4-Chloro-6,8-difluoro-2-methylquinoline
4-Chloro-6,8-difluoro-2-methylquinoline
Formel:
C₁₀H₆ClF₂N Molecular Weight: 213,611 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11501277 CAS-Nummer: 288151-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...