3-(Pentafluorothio)thiophenol
Lieferant: Apollo Scientific
|
Achtung
|
PC405648-1G
PC405648-250MG
APOSPC405648-1GEA
0
EUR
APOSPC405648-1G
APOSPC405648-250MG
3-(Pentafluorothio)thiophenol
3-(Pentafluorothio)thiophenol
Formel:
C₆H₅F₅S₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD27664885 CAS-Nummer: 1820618-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...