1-(3-Fluorophenyl)bicyclo[1.1.1]pentane

Lieferant: Apollo Scientific

PC405685-1G PC405685-250MG
APOSPC405685-1GEA 1660 EUR
APOSPC405685-1G APOSPC405685-250MG
1-(3-Fluorophenyl)bicyclo[1.1.1]pentane
1-(3-Fluorophenyl)bicyclo[1.1.1]pentane
Formel: C₁₁H₁₁F
Molecular Weight: 162,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27996102
CAS-Nummer: 1823878-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR