1-Fluoro-3-phenylbicyclo[1.1.1]pentane
Lieferant: Apollo Scientific
PC405691-1G
PC405691-250MG
APOSPC405691-1GEA
1910
EUR
APOSPC405691-1G
APOSPC405691-250MG
1-Fluoro-3-phenylbicyclo[1.1.1]pentane
1-Fluoro-3-phenylbicyclo[1.1.1]pentane
Formel:
C₁₁H₁₁F Molecular Weight: 162,203 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29037338 CAS-Nummer: 146038-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...