1-Fluoro-3-(4-iodophenyl)bicyclo[1.1.1]pentane

Lieferant: Apollo Scientific
Achtung

PC405692-1G PC405692-250MG
APOSPC405692-1GEA 3640 EUR
APOSPC405692-1G APOSPC405692-250MG
1-Fluoro-3-(4-iodophenyl)bicyclo[1.1.1]pentane
1-Fluoro-3-(4-iodophenyl)bicyclo[1.1.1]pentane
Formel: C₁₁H₁₀FI
Molecular Weight: 288,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037339
CAS-Nummer: 1934526-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR