(4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)phenyl)methanol

Lieferant: Apollo Scientific

PC405696-1G PC405696-250MG
APOSPC405696-1GEA 2410 EUR
APOSPC405696-1G APOSPC405696-250MG
(4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)phenyl)methanol
(4-(3-Fluorobicyclo[1.1.1]pentan-1-yl)phenyl)methanol
Formel: C₁₂H₁₃FO
Molecular Weight: 192.229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037343
CAS-Nummer: 1934767-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR