4-Chloro-6-trifluoromethoxyquinoline

Lieferant: Apollo Scientific

PC407013-1G PC407013-250MG
APOSPC407013-1GEA 260 EUR
APOSPC407013-1G APOSPC407013-250MG
4-Chloro-6-trifluoromethoxyquinoline
4-Chloro-6-trifluoromethoxyquinoline
Formel: C₁₀H₅ClF₃NO
Molecular Weight: 247.601 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684224
CAS-Nummer: 927800-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR