4-Chloro-7-trifluoromethylquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

PC407015-1G PC407015-5G
APOSPC407015-1GEA 0 EUR
APOSPC407015-1G APOSPC407015-5G
4-Chloro-7-trifluoromethylquinoline-3-carboxylic acid ethyl ester
4-Chloro-7-trifluoromethylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₉ClF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07700248
CAS-Nummer: 21168-42-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR