2'-Chloro-3'-fluoroacetophenone
Lieferant: Apollo Scientific
Synonyme:
1-(2-Chloro-3-fluorophenyl)ethanone
PC408052-1G
PC408052-5G
APOSPC408052-1GEA
60.9
EUR
APOSPC408052-1G
APOSPC408052-5G
2'-Chloro-3'-fluoroacetophenone
2'-Chloro-3'-fluoroacetophenone
Formel:
C₈H₆ClFO Molecular Weight: 172,584 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13194221 CAS-Nummer: 161957-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...