4,4,4-Trifluorocrotonoyl chloride 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC408119-25G
PC408119-5G
APOSPC408119-25GEA
915
EUR
APOSPC408119-25G
APOSPC408119-5G
4,4,4-Trifluorocrotonoyl chloride 98%
4,4,4-Trifluorocrotonoyl chloride
Formel:
C₄H₂ClF₃O Molecular Weight: 158,506 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092807 CAS-Nummer: 406-92-8 UN: 2920 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...