1,3-Diiodotetrafluorobenzene 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
m-Diiodoperfluorobenzene, 2,4,5,6-Tetrafluoro-1,3-diiodobenzene
PC408156-100G
APOSPC408156-100GEA
0
EUR
APOSPC408156-100G
1,3-Diiodotetrafluorobenzene 97%
1,3-Diiodotetrafluorobenzene
Formel:
C₆F₄I₂ Molecular Weight: 401,867 g/mol Siedepunkt: 140 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD16620566 CAS-Nummer: 67815-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...