1,4-Bis(fluorosulphonyloxy)octafluorobutane

Lieferant: Apollo Scientific
Gefahr

PC408186-1G PC408186-5G
APOSPC408186-1GEA 87 EUR
APOSPC408186-1G APOSPC408186-5G
1,4-Bis(fluorosulphonyloxy)octafluorobutane
1,4-Bis(fluorosulphonyloxy)octafluorobutane
Formel: C₄F₁₀O₆S₂
Molecular Weight: 398.153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00748183
CAS-Nummer: 78522-71-1
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR