2-Bromo-6-(trifluoromethoxy)benzoic acid 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC408236-1G
PC408236-5G
PC408236-10G
APOSPC408236-1GEA
55.1
EUR
APOSPC408236-1G
APOSPC408236-5G
APOSPC408236-10G
2-Bromo-6-(trifluoromethoxy)benzoic acid 99%
2-Bromo-6-(trifluoromethoxy)benzoic acid
Formel:
C₈H₄BrF₃O₃ Molecular Weight: 285,015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09266205 CAS-Nummer: 403646-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...