5-Bromo-2-(trifluoromethoxy)benzoic acid 99%

Lieferant: Apollo Scientific
Achtung

PC408237-1G PC408237-25G PC408237-5G
APOSPC408237-1GEA 43.5 EUR
APOSPC408237-1G APOSPC408237-25G APOSPC408237-5G
5-Bromo-2-(trifluoromethoxy)benzoic acid 99%
5-Bromo-2-(trifluoromethoxy)benzoic acid
Formel: C₈H₄BrF₃O₃
Molecular Weight: 285,015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203214
CAS-Nummer: 403646-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR