5-Nitro-2-(trifluoromethoxy)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC408278-10G PC408278-1G PC408278-5G
APOSPC408278-10GEA 0 EUR
APOSPC408278-10G APOSPC408278-1G APOSPC408278-5G
5-Nitro-2-(trifluoromethoxy)benzoic acid
5-Nitro-2-(trifluoromethoxy)benzoic acid
Formel: C₈H₄F₃NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD11847469
CAS-Nummer: 307989-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR