4-(Difluoromethyl)piperidine hydrochloride 97%
Lieferant: Apollo Scientific
PC408512-250MG
PC408512-1G
PC408512-5G
APOSPC408512-250MGEA
133
EUR
APOSPC408512-250MG
APOSPC408512-1G
APOSPC408512-5G
4-(Difluoromethyl)piperidine hydrochloride 97%
4-(Difluoromethyl)piperidine hydrochloride
Formel:
C₆H₁₁F₂N·ClH Molecular Weight: 171,616 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18711662 CAS-Nummer: 760958-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...