1-Bromo-2-(pentafluoroethyl)benzene 95%

Lieferant: Apollo Scientific

PC408835-1G PC408835-250MG PC408835-5G
APOSPC408835-1GEA 401 EUR
APOSPC408835-1G APOSPC408835-250MG APOSPC408835-5G
1-Bromo-2-(pentafluoroethyl)benzene 95%
1-Bromo-2-(pentafluoroethyl)benzene
Formel: C₈H₄BrF₅
Molecular Weight: 275,013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10000932
CAS-Nummer: 112754-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR