1-Bromo-3-(pentafluoroethyl)benzene

Lieferant: Apollo Scientific
Achtung

PC408836-1G PC408836-250MG
APOSPC408836-1GEA 261 EUR
APOSPC408836-1G APOSPC408836-250MG
1-Bromo-3-(pentafluoroethyl)benzene
1-Bromo-3-(pentafluoroethyl)benzene
Formel: C₈H₄BrF₅
Molecular Weight: 275.013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24386133
CAS-Nummer: 18739-03-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR