N-t-BOC-trans-4-Fluoro-L-prolinonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC409013-1G
PC409013-250MG
APOSPC409013-1GEA
619
EUR
APOSPC409013-1G
APOSPC409013-250MG
N-t-BOC-trans-4-Fluoro-L-prolinonitrile
N-t-BOC-trans-4-Fluoro-L-prolinonitrile
Formel:
C₁₀H₁₅FN₂O₂ Molecular Weight: 214.237 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458684 CAS-Nummer: 1330286-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...