4-Fluoro-4-Piperidinemethanol hydrochloride

Lieferant: Apollo Scientific

Synonyme: (4-Fluoropiperidin-4-yl)methanol hydrochloride

PC409057-1G PC409057-250MG
APOSPC409057-1GEA 163 EUR
APOSPC409057-1G APOSPC409057-250MG
4-Fluoro-4-Piperidinemethanol hydrochloride
4-Fluoro-4-Piperidinemethanol hydrochloride
Formel: C₆H₁₂FNO·ClH
Molecular Weight: 169,625 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19105116
CAS-Nummer: 1254115-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR