2-(3,3-Difluorocyclobutyl)acetonitrile

Lieferant: Apollo Scientific

PC411275-1G PC411275-500MG PC411275-5G
APOSPC411275-1GEA 655 EUR
APOSPC411275-1G APOSPC411275-500MG APOSPC411275-5G
2-(3,3-Difluorocyclobutyl)acetonitrile
2-(3,3-Difluorocyclobutyl)acetonitrile
Formel: C₆H₇F₂N
Molecular Weight: 131,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24856416
CAS-Nummer: 1219019-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR