3-(Pentafluoroethyl)-1H-pyrazol-5-ol
Lieferant: Apollo Scientific
|
Achtung
|
PC411320-1G
PC411320-5G
APOSPC411320-1GEA
48
EUR
APOSPC411320-1G
APOSPC411320-5G
3-(Pentafluoroethyl)-1H-pyrazol-5-ol
3-(Pentafluoroethyl)-1H-pyrazol-5-ol
Formel:
C₅H₃F₅N₂O Molecular Weight: 202,082 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04968776 CAS-Nummer: 173468-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...