4,4-Difluoro-1-(2-furyl)butane-1,3-dione

Lieferant: Apollo Scientific
Achtung

PC411330-1G PC411330-5G
APOSPC411330-1GEA 395 EUR
APOSPC411330-1G APOSPC411330-5G
4,4-Difluoro-1-(2-furyl)butane-1,3-dione
4,4-Difluoro-1-(2-furyl)butane-1,3-dione
Formel: C₈H₆F₂O₃
Molecular Weight: 188.128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03420652
CAS-Nummer: 480438-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR