5-(Cyclopropylcarbamoyl)-2-fluorophenylboronic acid

Lieferant: Apollo Scientific

PC412106-1G PC412106-250MG
APOSPC412106-1GEA 390 EUR
APOSPC412106-1G APOSPC412106-250MG
5-(Cyclopropylcarbamoyl)-2-fluorophenylboronic acid
5-(Cyclopropylcarbamoyl)-2-fluorophenylboronic acid
Formel: C₁₀H₁₁BFNO₃
Molecular Weight: 223,009 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11855834
CAS-Nummer: 874289-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR