2-Bromo-4,6-difluorophenylboronic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Bromo-4,6-difluorophenyl)boronic acid, 2,4-Difluoro-6-bromophenylboronic acid
PC412117-1G
PC412117-250MG
PC412117-5G
APOSPC412117-1GEA
224
EUR
APOSPC412117-1G
APOSPC412117-250MG
APOSPC412117-5G
2-Bromo-4,6-difluorophenylboronic acid 95%
2-Bromo-4,6-difluorophenylboronic acid
Formel:
C₆H₄BBrF₂O₂ Molecular Weight: 236,807 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09751350 CAS-Nummer: 1315339-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...