3-Bromo-4-fluorobenzeneboronic acid 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Bromo-4-fluorophenyl)boronic acid, 3-Bromo-4-fluorophenylboronic acid

PC412252-1G PC412252-250MG PC412252-5G
APOSPC412252-1GEA 213 EUR
APOSPC412252-1G APOSPC412252-250MG APOSPC412252-5G
3-Bromo-4-fluorobenzeneboronic acid 99%
3-Bromo-4-fluorobenzeneboronic acid
Formel: C₆H₅BBrFO₂
Molecular Weight: 218.816 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13195642
CAS-Nummer: 1092533-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR