4-(tert-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(t-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid
PC412272-1G
PC412272-5G
PC412272-10G
APOSPC412272-1GEA
120
EUR
APOSPC412272-1G
APOSPC412272-5G
APOSPC412272-10G
4-(tert-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid 98%
4-(tert-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid
Formel:
C₁₂H₁₉BF₂O₃Si Molecular Weight: 288,171 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18914502 CAS-Nummer: 870646-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...