3-Chloro-5-cyano-2-fluorophenylboronic acid, pinacol ester 96%
Lieferant: Apollo Scientific
PC412286-1G
PC412286-250MG
APOSPC412286-1GEA
1020
EUR
APOSPC412286-1G
APOSPC412286-250MG
3-Chloro-5-cyano-2-fluorophenylboronic acid, pinacol ester 96%
3-Chloro-5-cyano-2-fluorophenylboronic acid, pinacol ester
Formel:
C₁₃H₁₄BClFNO₂ Molecular Weight: 281,518 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15143597 CAS-Nummer: 1218790-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...