3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Chloro-2-fluoropyridne-5-boronic acid pinacol ester
PC412317-1G
PC412317-250MG
APOSPC412317-1GEA
295
EUR
APOSPC412317-1G
APOSPC412317-250MG
3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester
3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester
Formel:
C₁₁H₁₄BClFNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16995268 CAS-Nummer: 1220219-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...