2,5-Difluoropyridine-4-boronic acid

Lieferant: Apollo Scientific

Synonyme: (2,5-Difluoropyridin-4-yl)boronic acid

PC412380-1G PC412380-250MG
APOSPC412380-1GEA 194 EUR
APOSPC412380-1G APOSPC412380-250MG
2,5-Difluoropyridine-4-boronic acid
2,5-Difluoropyridine-4-boronic acid
Formel: C₅H₄BF₂NO₂
Molecular Weight: 158.899 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16556243
CAS-Nummer: 1263375-23-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR