2-Fluoro-6-hydroxyphenylboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Fluoro-6-hydroxyphenyl)boronic acid
PC412426-1G
PC412426-250MG
PC412426-5G
PC412426-10G
APOSPC412426-1GEA
24.8
EUR
APOSPC412426-1G
APOSPC412426-250MG
APOSPC412426-5G
APOSPC412426-10G
2-Fluoro-6-hydroxyphenylboronic acid
2-Fluoro-6-hydroxyphenylboronic acid
Formel:
C₆H₆BFO₃ Molecular Weight: 155,919 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD17214242 CAS-Nummer: 1256345-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...