5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 5-Fluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester
PC412439-1G
PC412439-250MG
PC412439-5G
APOSPC412439-1GEA
122
EUR
APOSPC412439-1G
APOSPC412439-250MG
APOSPC412439-5G
5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester 97%
5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester
Formel:
C₁₄H₁₈BFO₄ Molecular Weight: 280.10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16996304 CAS-Nummer: 1400976-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...