1-(Methoxymethyl)-3-(trifluoromethyl)pyrazole-4-boronic acid
Lieferant: Apollo Scientific
Synonyme:
(1-(Methoxymethyl)-3-(trifluoromethyl)-1H-pyrazol-4-yl)boronic acid
PC412510-250MG
PC412510-100MG
APOSPC412510-250MGEA
771
EUR
APOSPC412510-250MG
APOSPC412510-100MG
1-(Methoxymethyl)-3-(trifluoromethyl)pyrazole-4-boronic acid
1-(Methoxymethyl)-3-(trifluoromethyl)pyrazole-4-boronic acid
Formel:
C₆H₈BF₃N₂O₃ Molecular Weight: 223,946 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15143477 CAS-Nummer: 1218790-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...